Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Chloride Hexahydrate, (Crystalline),MP Biomedicals

Catalog No. ICN19471390 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19471390 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19471390 Supplier MP Biomedicals, Inc Supplier No. 0219471390
Add to Cart
Add to Cart

Nickel chloride hexahydrate optimized for use as a cell culture reagent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-20-0
Molecular Formula NiCl2 · 6H2O
Molecular Weight (g/mol) 237.71

Specifications

Physical Form Crystalline
Quantity 500 g
Formula Weight 237.7
Grade Cell Culture
Chemical Name or Material Nickel Chloride Hexahydrate
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.