Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Chloride Hexahydrate, (Crystalline),MP Biomedicals

Catalog No. ICN15582591 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15582591 1 kg
ICN19471380 100 g
ICN15582590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15582591 Supplier MP Biomedicals, Inc Supplier No. 0215582591
Add to Cart
Add to Cart

Cell Culture Reagent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-20-0
Molecular Formula NiCl2 · 6H2O

Specifications

Physical Form Crystalline
Quantity 1 kg
Formula Weight 237.7
Chemical Name or Material Nickel Chloride Hexahydrate
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.