Learn More
Nickel(II) 2,4-pentanedionate hydrate, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Boiling Point | 220°C (11mmHg) |
| Melting Point | ∼230°C (decomposition) |
| Physical Form | Powder |
| Quantity | 250 g |
| Beilstein | 4157970 |
| Merck Index | 14,6500 |
| Formula Weight | 256.91 (Anhydrous) |
| Chemical Name or Material | Nickel(II) 2,4-pentanedionate hydrate |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H351 Suspected of causing cancer. |
| Precautionary Statement | GHS P Statement P281-P201-P202-P308+P313-P405-P501a Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. IF exposed or concerned: Get medical advice/attention. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| RTECSNumber | SA2100000 |
| TSCA | TSCA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.