Learn More
Nickel(II) acetate tetrahydrate, 98+%
Description
Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.
Solubility
Soluble in water and methanol. Insoluble in diethyl ether and n-octanol.
Notes
Hygroscopic. Incompatible with oxidizing agents, strong acids and strong bases.
Specifications
Specifications
| Melting Point | (decomposition) |
| Density | 1.744 |
| Odor | Light |
| Quantity | 2500 g |
| UN Number | UN3077 |
| Beilstein | 3730764 |
| Merck Index | 14,6499 |
| Solubility Information | Soluble in water and methanol. Insoluble in diethyl ether and n-octanol. |
| Formula Weight | 248.84 (176.79 Anhydrous) |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H317-H334-H335-H341-H350i-H360D-H372 |
| Hazard Statement | GHS H Statement H334-H350-H360-H372-H341-H302-H332-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 206-761-7 |
| RTECSNumber | QR6126000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.