Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) Benzenesulfonate Hexahydrate 98.0+%, TCI America™
SDP

Catalog No. B003425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B003425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B003425G Supplier TCI America Supplier No. B003425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39819-65-3
Molecular Formula C12H10NiO6S2
Molecular Weight (g/mol) 373.019
MDL Number MFCD00070595
InChI Key IBVWKDVFDAWRFU-UHFFFAOYSA-L
Synonym Benzenesulfonic Acid Nickel(II) Salt
PubChem CID 14325094
IUPAC Name benzenesulfonate;nickel(2+)
SMILES C1=CC=C(C=C1)S(=O)(=O)[O-].C1=CC=C(C=C1)S(=O)(=O)[O-].[Ni+2]

Specifications

Quantity 25 g
UN Number 3077
Formula Weight 373.02
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nickel(II) Benzenesulfonate Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.