Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) chloride hexahydrate, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 5313118

Quantity 50g

 View more versions of this product

Catalog No. AA5313118


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7791-20-0
99.95% (metals basis)
237.71
Nickel(II) chloride hexahydrate
Crystalline
Cl2Ni·6H2O
MFCD00149809

Specifications

7791-20-0
99.95% (metals basis)
237.71
1.92g/cm3 at 20°C
Green
Nickel(II) chloride hexahydrate
Crystalline
Cl2Ni·6H2O
MFCD00149809
Odorless
50g

Safety and Handling

SDS & Safety Documents

H301-H331-H315-H317-H334-H350-H360-H372-H341-H400-H410: Toxic if swallowed. Toxic if inhaled. Causes skin irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 231-743-0

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit