Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel (II) Chloride Hexahydrate, ACS Reagent, Reagents

Catalog No. 01337882
Change view
Click to view available options
Quantity:
125 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-882 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-882 Supplier Reagents Holdings Llc Supplier No. C1255500125D2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nickel (II) Chloride Hexahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2H12NiO6
Molecular Weight (g/mol) 237.68
InChI Key LAIZPRYFQUWUBN-UHFFFAOYSA-L
Synonym Nickel(II) Chloride Hexahydrate, Nickelous Chloride Hexahydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ni++]

Specifications

Boiling Point 973°C
Color Green
Physical Form Solid
Quantity 125 g
IUPAC Name nickel(2+) hexahydrate dichloride
Percent Purity 98%
Grade ACS
Packaging Glass Bottle
Specific Gravity 3.55
Chemical Name or Material Nickel (II) Chloride Hexahydrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.