Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(Ii) Chloride Hexahydrate, ReagentPlus™, Solstice

Catalog No. 60045349 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-349 2 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-349 Supplier Solstice Supplier No. 2233872KG

missing translation for 'validMainframeMsg'

Reagent Grade

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-20-0
MDL Number MFCD00149809

Specifications

Quantity 2 kg
Linear Formula NiCl2 · 6H2O
UN Number UN3288
Formula Weight 237.69g/mol
Grade ReagentPlus
Packaging Poly Bottle
Chemical Name or Material Nickel(Ii) Chloride Hexahydrate
Hazard Statement P201-P261-P273-P280-P284-P301 + P310 + P330
Precautionary Statement H301 + H331-H315-H317-H334-H341-H350i-H360D-H372-H410
DOTInformation Class 6.1 - PG 3 - EHS - RQ - Toxic solid, inorganic, n.o.s., HI: all
RTECSNumber QR6480000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.