Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Nickel (II) Nitrate, Hexahydrate
SCI_ED

Catalog No. S25443 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25443 100 g Reagent Grade
S25443A 500 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25443

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 13478-00-7
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
Synonym nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
PubChem CID 61630
IUPAC Name nickel(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Green
Quantity 100 g
Assay Percent Range 99%
Linear Formula Ni(NO3)2·6H2O
Formula Weight 290.82
Grade Reagent
Packaging Reagent Grade
Density 2.05g/mL
Chemical Name or Material Nickel (II) Nitrate, Hexahydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
DOTInformation DOT Class 5.1, III, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.