Learn More
Nickel(II) Sulfate Hexahydrate, Reagent Grade, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- ≥95% for general laboratory use.
Specifications
Specifications
| Melting Point | 53°C |
| Color | Green |
| pH | 4 to 6 (at 20°C) |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Linear Formula | NiSO4 · 6H2O |
| UN Number | UN3077 |
| Formula Weight | 262.85g/mol |
| Odor | Odorless |
| Packaging | Poly bottle |
| Show More |
Safety and Handling
| Hazard Statement | P201-P261-P273-P280-P284-P304 + P340 + P312 |
| DOTInformation | 9/PGIII |
| RTECSNumber | 232-104-9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.