Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Iron Copper Molybdenum foil, 0.125mm (0.005 in.) thick, annealed (Mumetal™)

Catalog No. AA43399FL
Change view
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43399FL 75 x 75 mm
AA43399GT 150 x 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43399FL Supplier Thermo Scientific Chemicals Supplier No. 043399FL
Add to Cart

Magnetic shielding alloy

Nickel Iron Copper Molybdenum foil is used as a magnetic shielding alloy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel Iron Copper Molybdenum foil
Name Note 0.125mm (0.005 in.) thick, Annealed (Mumetal™)
Avg. Mol. Wt. or Mol. Wt. Range Ni:Fe:Cu:Mo; 77:14:5:4 wt%
Quantity 75 x 75 mm
Form Foil
MDL Number MFCD02684505
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.