Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Nitrate Hexahydrate, ACS, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18611973 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-973 125 g Amber Glass Bottle
18-611-972 25 g Amber Glass Bottle
18-611-974 500 g Amber Glass Bottle
18-611-975 2.5 kg Amber Glass Bottle
18-611-976 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-611-973 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N1267125GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13478-00-7
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
IUPAC Name nickel(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98 to 102%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.