Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Nickel-plated Spatula

Catalog No. 14127268
Change view
Click to view available options
Length (Metric):
150 mm
225 mm
Shape:
Hayman-type end with flat spoon
Spoon end with flat handle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Shape
14-127-268 150 mm Hayman-type end with flat spoon
14-127-267 225 mm Spoon end with flat handle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-127-268 Supplier Heathrow Scientific™ Supplier No. HS15906
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spoons out powder and granular materials. Heathrow Scientific™ Nickel-plated Spatula also contains stainless steel wire for durability.

  • Made from nickel-plated, stainless steel wire
  • Autoclavable

Specifications

Disposable Reusable
Product Type Spoon
Finish Glazed
Length (Metric) Handle 30 mm
Material Stainless Steel
Handle Type Hayman
Length (Metric) 150 mm
Autoclavable Autoclavable
Shape Hayman-type end with flat spoon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.