Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Sulfate, Hexahydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604802 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-802 25 g Poly Bottle
18-604-803 125 g Poly Bottle
18-604-804 500 g Poly Bottle
18-604-806 2.5 kg Poly Pail
18-604-805 6 x 500 g Poly Bottle
18-604-807 12 kg Poly Pail
18-604-808 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-604-802 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N107025GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

N1070, 10101-97-0, NiSO4·6H2O

Spectrum™ Chemical Nickel Sulfate, Hexahydrate, Crystal, Reagent, ACS is an inorganic compound that appears as a blue colored salt. Nickel Sulfate is also considered highly soluble and commonly used for electroplating of nickel. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.84
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
IUPAC Name nickel(2+) hexahydrate sulfate
SMILES O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98 to 102%
Grade ACS
Packaging Poly Bottle
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.