Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel tungsten oxide, 99.9% (metals basis excluding Co), Co <100ppm

Catalog No. AA4182614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4182614 25 g
AA4182622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4182614 Supplier Thermo Scientific Chemicals Supplier No. 04182614
Add to Cart

CAS: 14177-51-6 | NiO4W | 306.529 g/mol

Nickel tungsten oxide is used as photo catalysts, sensors and humidity dielectric resonators. It is considered as a new cathode material for asymmetric super capacitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14177-51-6
Molecular Formula NiO4W
Molecular Weight (g/mol) 306.529
MDL Number MFCD00016268
InChI Key QLTKZXWDJGMCAR-UHFFFAOYSA-N
Synonym nickel tungstate, nickel tungsten tetraoxide, nickel tungsten oxide niwo4, nickel 2+ dioxido dioxo tungsten, nickel 2+ ion tungstate, dioxido dioxo tungsten; nickel 2+
PubChem CID 84240
IUPAC Name dioxido(dioxo)tungsten;nickel(2+)
SMILES [O-][W](=O)(=O)[O-].[Ni+2]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 306.53
Percent Purity 99.9%
Odor Odorless
Assay (metals basis excluding Co), Co <100ppm
Chemical Name or Material Nickel tungsten oxide
Hazard Category H317-H350i-H372
Hazard Statement GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 238-032-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.