Learn More
Eisco™ Nickle Plated Sampling Tong
SCI_ED
Catalog No. S41736 Shop All Eisco Products
Change view
Click to view available options
Length (Metric):
20 cm
Catalog No. Length (Metric)
S41736 20 cm
1 options
Catalog No. S41736 Supplier Eisco™ Supplier No. CH0736
Add to Cart
Add to Cart
Nickle Plated Sampling Tong splits in half vertically which is useful for collecting hot samples of thick materials. Designed to handle mercury safely. Scalloped spoon design, half scallop splits into a quarter per side.
  • Designed to handle globules of mercury safely.
  • Scalloped spoon design splits into a quarter per side.
Warranty and Services
1 year limited manufacturer warranty
Specifications
Product Type Sampling Tongs
Material Nickle-plated Metal
Length (Metric) 20 cm
Quantity 1
Includes Tongs
For Use With (Application) Collecting samples