Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niclosamide, 97+%, Thermo Scientific Chemicals

Catalog No. AC461841000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461841000 100 g
AC461840250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461841000 Supplier Thermo Scientific Chemicals Supplier No. 461841000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-65-7
Molecular Formula C13H8Cl2N2O4
Molecular Weight (g/mol) 327.12
MDL Number MFCD00057597
InChI Key RJMUSRYZPJIFPJ-UHFFFAOYSA-N
Synonym niclosamide, 5-chloro-n-2-chloro-4-nitrophenyl-2-hydroxybenzamide, niclocide, bayluscid, phenasal, tredemine, fenasal, yomesan, dichlosale, helmiantin
PubChem CID 4477
IUPAC Name 5-chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide
SMILES OC1=CC=C(Cl)C=C1C(=O)NC1=CC=C(C=C1Cl)[N+]([O-])=O

Specifications

Melting Point 225.0°C to 230.0°C
Assay Percent Range 97% min. (HPLC)
Quantity 100 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: sparingly soluble in ethanol, chloroform, ether
Formula Weight 327.12
Percent Purity 97+%
Chemical Name or Material Niclosamide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 200-056-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.