accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClONHClNOO

CAS RN 50-65-7

IUPAC Name: 5-chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide
Synonyms: niclosamide 氯硝柳胺 niclocide
Molecular Weight (g/mol): 327.12
Molecular Formula: C13H8Cl2N2O4
InChi Key: RJMUSRYZPJIFPJ-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1C(=O)NC1=CC=C(C=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (5)
Grade 
  • (5)
  • (2)
Physical Form 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (5)
  • (2)
  • (5)
  • (2)