Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niclosamide, Anhydrous, BP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18604812 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-812 25 g Amber Glass Bottle
18-604-809 100 g Amber Glass Bottle
18-604-813 500 g Amber Glass Bottle
18-604-810 1 kg Amber Glass Bottle
18-604-811 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-812 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N300925GM
Add to Cart
Add to Cart

N3009, 50-65-7, C13H8Cl2N2O4

Spectrum™ Chemical Niclosamide, Anhydrous, BP is a medication used to treat tapeworm infestations. All Spectrum Chemical BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-65-7
Molecular Formula C13H8Cl2N2O4
Molecular Weight (g/mol) 327.12
MDL Number MFCD00057597
InChI Key RJMUSRYZPJIFPJ-UHFFFAOYSA-N
IUPAC Name 5-chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide
SMILES OC1=CC=C(Cl)C=C1C(=O)NC1=CC=C(C=C1Cl)[N+]([O-])=O

Specifications

CAS Max % 1
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 101%
Grade BP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.