Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niclosamide, MP Biomedicals™

Catalog No. MP215582783 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215582783 250 g
MP215582780 100 g
MP215582750 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215582783 Supplier MP Biomedicals, Inc Supplier No. 0215582783
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Niclosamide ≥98%, Greenish-Yellow Powder

  • It is used to study the Wnt/Frizzled-1 signaling pathway.
  • It is used to inhibit the transcription and DNA binding of NF-κB in AML cells and to induce apoptosis.
  • It is effective against cestodes that infect humans.
  • Niclosamide uncouples oxidative phosphorylation in the tapeworm.
  • It inhibits mitochondrial oxidative phosphorylation of parasitic helminths.
  • It inhibits the NF-κB pathway in acute myelogenous leukemia (AML) cells.
  • Niclosamide inhibits the transcription and DNA binding of NF-κB.
  • It blocks tumor necrosis factor-induced IκBα phosphorylation, translocation of p65, and expression of NF-κB– regulated genes in AML cells. It increases ROS levels to induce apoptosis in AML cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-65-7
Molecular Formula C13H8Cl2N2O4
Molecular Weight (g/mol) 327.12
MDL Number MFCD00057597
InChI Key RJMUSRYZPJIFPJ-UHFFFAOYSA-N
Synonym 2´,5-Dichloro-4-nitrosalicylanilide
IUPAC Name 5-chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide
SMILES OC1=CC=C(Cl)C=C1C(=O)NC1=CC=C(C=C1Cl)[N+]([O-])=O

Specifications

Melting Point 225°C to 233°C
Color Green-yellow
Loss on Drying ≤0.5%
Quantity 250 g
UN Number UN3077
Beilstein 2820605
Solubility Information Soluble in methanol:acetone [1:1] (50mg/mL).
Vapor Pressure Very low at room temp
RTECSNumber VN8400000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.