Learn More
Apexbio Technology LLC PIK-294 900185-02-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B21885.1
Encompass_Preferred
PIK-294 (CAS 900185-02-6) is a potent and highly selective inhibitor of the class I phosphoinositide 3-kinase (PI3K) isoform p110 with an IC50 of 10 nM It exhibits substantially reduced activity against PI3K PI3K and PI3K demonstrating a 1 000-fold 49-fold and 16-fold lower potency respectively Biochemical studies indicate that PIK-294 interacts with a high-affinity pocket in the ATP-binding site contributing to its inhibitory properties in vitro Used as a chemical probe PIK-294 aids in delineating isoform-specific PI3K functions such as the distinct roles of PI3K isoforms in insulin signaling and cellular differentiation pathways No clinical studies have been reported
Catalog No. 50-000-89353
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.