Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Nicotinamide, 99%

Catalog No. AC128270025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC128270025 2.5 kg Plastic bottle
AC128271000 100 g Plastic bottle
AC128275000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC128270025 Supplier Thermo Scientific Chemicals Supplier No. 128270025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-92-0 | C6H6N2O | 122.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00006395
InChI Key DFPAKSUCGFBDDF-UHFFFAOYSA-N
Synonym nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide
PubChem CID 936
ChEBI CHEBI:17154
IUPAC Name pyridine-3-carboxamide
SMILES C1=CC(=CN=C1)C(=O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.0°C to 131.0°C
Color White
pH 6.0 to 7.5 (5% soln. at 20°C)
Boiling Point 150.0°C to 160.0°C
Flash Point >150°C
Loss on Drying 0.5% max. (4 hrs) (silica gel)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 22, 40
Merck Index 15, 6608
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: 1g/1.5mL alcohol,1g/10mL glycerol
Formula Weight 122.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Nicotinamide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 202-713-4
RTECSNumber QS3675000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.