Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Nicotinamide Adenine Dinucleotide, MP Biomedicals™

Catalog No. MP210031905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210031905 5 g
MP210031925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP210031905 Supplier MP Biomedicals, Inc Supplier No. 0210031905
Add to Cart
Add to Cart

Serves as a coenzyme of the dehydrogenases. β-Nicotinamide Adenine Dinucleotide, MP Biomedicals is one of the biologically active forms of nicotinic acid that occurs in living cells primarily in the oxidized state. It is a major electron acceptor mol

  • Serves as a coenzyme of the dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols
  • Usually acts as a hydrogen acceptor, forming NADH, which then serves as a hydrogen donor in the respiratory chain
  • Acts as a coenzyme for hydride-transfer enzymes
  • Used as a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins
  • Recently reported to be a P2Y11 agonist
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-84-9
Molecular Formula C21H27N7O14P2
Molecular Weight (g/mol) 663.43
MDL Number MFCD00150377
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
Synonym nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
PubChem CID 15938971
ChEBI CHEBI:57540
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

pH 2 to 4 (1% aq. soln.)
Quantity 5 g
Solubility Information Soluble in water (200mg/mL)
Formula Weight 663.40 (anhydrous)
Percent Purity ≥93%
Physical Form Powder
Recommended Storage −20°C, desiccate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.