Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ β-Nicotinamide Adenine Dinucleotide, Oxidized, MP Biomedicals™

Catalog No. MP021600471 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021600471 100 mg
MP021600472 250 mg
MP021600475 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP021600471 Supplier MP Biomedicals™ Supplier No. 02160047.1
Add to Cart
Add to Cart

Necessary for the catalytic reaction of certain enzymes. β-Nicotinamide Adenine Dinucleotide, Oxidized, MP Biomedicals is a pyridine nucleotide and biologically active form of nicotinic acid. β-NAD functions as an electron acceptor and occurs in

Many metabolites and enzymes of biological interest are present in tissues at low concentrations. With the use of β-NAD as a catalyst intermediate and several enzymes in a multistep system, known as enzyme cycling, much greater sensitivity for detection of these components is achieved. The reduced form, β-NADH, is fluorescent whereas β-NAD is not. This difference in fluorescence provides a sensitive fluorescent measurement of the oxidized or reduced pyridine nucleotides at concentrations down to 10-7 M.

  • Serves as a coenzyme of the dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols
  • NAD usually acts as an electron acceptor, acting as a carrier for hydride ion, forming b-NADH which then serves as a hydrogen donor in the respiratory chain
    • Hydride ion is enzymatically removed from a substrate molecule by the action of dehydrogenases such as malic dehydrogenase and lactic dehydrogenase
    • Such enzymes catalyze the reversible transfer of a hydride ion from malate or lactate to b-NAD to form the reduced product, b-NADH
    • Unlike b-NAD which has no absorbance at 340nm, b-NADH absorbs at 340nm (EmM = 6.22)
    • The increase in absorbance at 340nm with the formation of b-NADH is the basis for measurement of activity of many enzymes
TRUSTED_SUSTAINABILITY

Specifications

Color White
Purity ≥99%
pH 2 to 4 (1% aq soln)
Content And Storage -20°C, Store Desiccated
Quantity 100 mg
Molecular Weight (g/mol) 663.4 (anhyd.)
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.