Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Nicotinamide adenine dinucleotide phosphate, disodium salt, 98-100%, MP Biomedicals™

Catalog No. ICN10116690 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
500 mg
1 g
5 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10116690 500 mg
ICN10116650 50 mg
ICN10116680 100 mg
ICN10116683 250 mg
ICN10116691 1 g
ICN10116694 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN10116690 Supplier MP Biomedicals, Inc Supplier No. 0210116690
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24292-60-2
Molecular Formula C21H26N7Na2O17P3
Molecular Weight (g/mol) 787.37
MDL Number MFCD00065390
InChI Key WSDDJLMGYRLUKR-MWLGWILCNA-L
Synonym nadp disodium salt, beta-nicotinamide adenine dinucleotide phosphate disodium salt
PubChem CID 131851653
IUPAC Name [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
SMILES [Na+].[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Color White
Assay Percent Range 98-100%
Quantity 500 mg
Formula Weight 787.4
Physical Form Powder
Chemical Name or Material β-Nicotinamide adenine dinucleotide phosphate, disodium salt and monopotassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.