Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 95%, MP Biomedicals™

Catalog No. ICN15174250 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
250 mg
500 mg
1 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15174250 50 mg
ICN15174225 25 mg
ICN15174280 100 mg
ICN15174283 250 mg
ICN15174290 500 mg
ICN15174291 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN15174250 Supplier MP Biomedicals, Inc Supplier No. 0215174250
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2646-71-1
Molecular Formula C21H26N7Na4O17P3
Molecular Weight (g/mol) 833.35
MDL Number MFCD10567216
InChI Key WYWWVJHQDVCHKF-NRTBITFTNA-J
Synonym nadph tetrasodium salt, nadph, tetrasodium salt, coenzyme ii tetrasodium salt reduced form
PubChem CID 131673986
SMILES [Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Color White
Quantity 50 mg
IUPAC Name [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
Formula Weight 833.4
Percent Purity 0.95
Physical Form Powder
Chemical Name or Material β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.