Learn More
Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 100.0% | 290.70 g/mol | C11H15ClN2O5 | 100MG
Supplier: Medchemexpress LLC HY123033AR100MG
Nicotinamide riboside chloride is the chloride salt of nicotinamide riboside, an NAD+ precursor used in research to raise cellular NAD+ levels and investigate sirtuin-related metabolic and neuroprotective pathways. This analytical standard is supplied as a solid, high-purity compound intended for research and analytical applications.
- High purity suitable for analytical and research use.
- Solid, white to off-white appearance for easy handling.
- Molecular formula C11H15ClN2O5; molecular weight 290.70 g/mol.
- Available in small mg-scale packages for laboratory experiments.
- Intended for research use only.