Learn More
Medchemexpress LLC Nicotinamide riboside malate | 2415659-01-5 | MFCD34549739 | 98.0% | 388.33 g/mol | C15H20N2O10 | 100MG
SDP
Supplier:  Medchemexpress LLC HY123033C100MG
Encompass_Preferred
Nicotinamide riboside malate is a research-grade NAD+ precursor provided as a stable powder for biochemical, cellular, and in vivo studies. It elevates cellular NAD+ levels and is used to probe sirtuin-mediated metabolic and neuroprotective pathways; reported molecular weight is 388.33 g/mol and CAS 2415659-01-5.
  • Elevates cellular NAD+ levels for metabolic studies.
  • Activates sirtuin pathways, including SIRT1 and SIRT3.
  • Suitable for biochemical, cellular, and in vivo assays.
  • High purity for research applications (98.0%).
  • Available in small research pack sizes, such as 100 mg.
Catalog No. 50-002-35745