Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic acid, 99%

Catalog No. AAA126830I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA126830I 5000 g
AAA1268330 250 g
AAA126830B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA126830I Supplier Thermo Scientific Chemicals Supplier No. A126830I
Add to Cart

CAS: 59-67-6 | C6H5NO2 | 123.11 g/mol

Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP.Nicotinic acid is used as an antidyslipidemic agent. It is also used to improve all lipoproteins well above the vitamin requirement and lowers the total cholesterol, bad LDL-cholesterol, and triglyceride levels. Further, it is used as a food additive in food industries, cosmetics and pharmaceuticals. In addition to this, it is used in medication, nutrient, enriched flours and in electroplating baths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP.Nicotinic acid is used as an antidyslipidemic agent. It is also used to improve all lipoproteins well above the vitamin requirement and lowers the total cholesterol, ″bad″ LDL-cholesterol, and triglyceride levels. Further, it is used as a food additive in food industries, cosmetics and pharmaceuticals. In addition to this, it is used in medication, nutrient, enriched flours and in electroplating baths.

Solubility
Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006391
InChI Key PVNIIMVLHYAWGP-UHFFFAOYSA-N
Synonym nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
PubChem CID 938
ChEBI CHEBI:15940
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1

Specifications

Melting Point 236°C to 239°C
Density 1.4
pH 2.7
Flash Point 293°C (559°F)
Odor Odorless
Assay Percent Range 99%
Quantity 5000 g
Beilstein 109591
Merck Index 14,6525
Solubility Information Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents.
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material Nicotinic acid
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 200-441-0
RTECSNumber QT0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.