Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIDA 41020, R&D Systems™
SDP

Catalog No. 392150 Shop All Novus Products
Change view
Click to view available options
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
39-215-0 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-215-0 Supplier R&D Systems Supplier No. 3921/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. High affinity CB1 antagonist

High affinity CB1 receptor antagonist (Ki = 4.1 nM). Exhibits significantly reduced lipophilicity compared to other CB1 antagonists.

Specifications

CAS 502486-89-7
Molecular Formula C23H24Cl2N4O2
Quantity 50 mg
Synonym unii-83e62l67id, 1-2,4-dichlorophenyl-5-4-methoxyphenyl-4-methyl-n-1-piperidinyl-1h-pyrazole-3-carboxamide, n-piperidin-1-yl-5-4-methoxyphenyl-1-2,4-dichlorophenyl-4-methyl-1h-pyrazole-3-carboxamide, nida hplc , solid, 1-2,4-dichloro-phenyl-5-4-methoxy-phenyl-4-methyl-1h-pyrazole-3-carboxylic acid piperidin-1-ylamide, 1-2,4-dichlorophenyl-5-4-methoxyphenyl-4-methyl-n-piperidin-1-yl pyrazole-3-carboxamide, 1h-pyrazole-3-carboxamide, 1-2,4-dichlorophenyl-5-4-methoxyphenyl-4-methyl-n-1-piperidinyl
InChI Key KWDBQJRWPWTGPF-UHFFFAOYSA-N
SMILES CC1=C(N(N=C1C(=O)NN2CCCCC2)C3=C(C=C(C=C3)Cl)Cl)C4=CC=C(C=C4)OC
IUPAC Name 1-(2,4-dichlorophenyl)-5-(4-methoxyphenyl)-4-methyl-N-piperidin-1-ylpyrazole-3-carboxamide
Molecular Weight (g/mol) 459.371
PubChem CID 11037861
Formula Weight 459.37
Percent Purity >99%
Product Line Tocris
Chemical Name or Material NIDA 41020
Show More Show Less
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.