Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIF1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501558473
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8473 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8473 Supplier Bethyl Laboratories, Inc Supplier No. A300798AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Component or associated component of some histone methyltransferase complexes may regulate transcription through recruitment of those complexes on gene promoters. Enhances ligand-dependent transcriptional activation by nuclear hormone receptors. Plays an important role in neural progenitor cell proliferation and self-renewal through the regulation of specific genes involved brain development, including REST. Also controls the expression of genes involved in somatic development and regulates, for instance, lymphoblast proliferation.

Specifications

Antigen NIF1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ZNF335
Gene Accession No. A2A5K6, Q9H4Z2
Gene Alias MCPH10; NIF-1; NIF1; NIF2; zinc finger protein 335; NRC-interacting factor 1; zinc-finger/leucine-zipper co-transducer NIF1
Gene Symbols Zfp335, ZNF335
Host Species Rabbit
Immunogen Between 1300 and C-term
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329559, 63925
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.