Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Nifedipine, 98+%

Catalog No. AC329270050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC329270050 5 g Glass bottle
AC329270010 1 g Glass bottle
AC329270250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC329270050 Supplier Thermo Scientific Chemicals Supplier No. 329270050
  • $177.00
    $142.65 / Each of 1

    Save $34.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 21829-25-4 | C17H18N2O6 | 346.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21829-25-4
Molecular Formula C17H18N2O6
Molecular Weight (g/mol) 346.34
MDL Number MFCD00057326
InChI Key HYIMSNHJOBLJNT-UHFFFAOYSA-N
Synonym nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord
PubChem CID 4485
ChEBI CHEBI:7565
IUPAC Name dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC

Specifications

Melting Point 171.0°C to 175.0°C
Color White to Yellow
Ignition Residue 0.1% max.
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Beilstein 22, V,4, 268
Merck Index 15, 6613
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone,chloroform,benzene,dioxane,,methanol,ethanol
Formula Weight 346.3
Percent Purity 98.0 to 102.0% (on dried substance)
Physical Form Powder
Chemical Name or Material Nifedipine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 244-598-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.