Learn More
Description
Nifedipine USP is a yellow powder. It is affected by exposure to light.
Specifications
Specifications
| Chemical Name or Material | Nifedipine USP |
| Packaging | Jar, 1000mL, White, HDPE, Round, 89-400, TE Pano Neck |
| CAS | 21829-25-4 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 346.3372 |
| Quantity | 500 g |
| Percent Purity | 98 to 102 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
