Learn More
Invitrogen™ NIFK Monoclonal Antibody (18E148)

Description
Suggested positive control: antigen standard for MKI67IP (transient overexpression lysate), HeLa or Ramos.
Specifications
Specifications
| Antigen | NIFK |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | '18E148 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | Nifk |
| Gene Accession No. | Q9BYG3 |
| Gene Alias | AI852665; C130020J04Rik; hNIFK; Ki67ip; MKI67 (FHA domain) interacting nucleolar phosphoprotein; MKI67 FHA domain-interacting nucleolar phosphoprotein; MKI67IP; mNIFK; NIFK; NOPP34; nucleolar phosphoprotein Nopp34; nucleolar protein interacting with the FHA domain of MKI67; nucleolar protein interacting with the FHA domain of pKI-67; spermatogenesis-related protein (Srp) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.