Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIFK Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675521
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-521 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-521 Supplier Invitrogen™ Supplier No. PA141392
  • $587.00
    $487.70 / Each of 1

    Save $99.30

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: antigen standard for MKI67IP (transient overexpression lysate), NIH-3T3 whole cell lysate.

NIFK, also known as nucleolar protein interacting with the forkhead homolog protein, is a gene associated with several cancer types and is considered a potential prognostic biomarker. This gene functions within the nucleolus and plays a critical role in cellular proliferation, metastasis, and invasiveness, particularly noted in hepatocellular carcinoma (HCC) and colorectal cancer. NIFK expression is regulated via several pathways, including the MYC proto-oncogene pathway, and its silencing can lead to decreased fatty acid synthesis in colorectal cancer cells. Furthermore, it's involved in the regulation of innate immunity through pathways such as NF-kB in specific cancer cells. Due to its involvement in cancer progression, NIFK is being explored as a therapeutic target and holds promise for advancing precision medicine strategies across various cancer treatments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIFK
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Nifk
Gene Accession No. Q91VE6, Q9BYG3
Gene Alias AI852665; C130020J04Rik; hNIFK; Ki67ip; MKI67 (FHA domain) interacting nucleolar phosphoprotein; MKI67 FHA domain-interacting nucleolar phosphoprotein; MKI67IP; mNIFK; NIFK; NOPP34; nucleolar phosphoprotein Nopp34; nucleolar protein interacting with the FHA domain of MKI67; nucleolar protein interacting with the FHA domain of pKI-67; spermatogenesis-related protein (Srp)
Gene Symbols Nifk
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 75-90 of mouse NIFK.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67949, 84365
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.