Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niflumic acid, Tocris Bioscience™
SDP

Catalog No. 411250
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-125-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 41-125-0 Supplier R&D Systems Supplier No. 4112/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cyclooxygenase-2 (COX-2) inhibitor; GPR35 agonist

Nonsteroidal anti-inflammatory drug (NSAID); selectively inhibits COX-2. Also activates human TRPA1 inducibly expressed in HEK293 cells; displays agonist activity at GPR35.

Chemical Identifiers

CAS 4394-00-7
Molecular Formula C13H9F3N2O2
Molecular Weight (g/mol) 282.22
MDL Number MFCD00010569
InChI Key JZFPYUNJRRFVQU-UHFFFAOYSA-N
Synonym niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french
PubChem CID 4488
ChEBI CHEBI:34888
IUPAC Name 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F

Specifications

Quantity 50 mg
Formula Weight 282.22
Percent Purity >98%
Chemical Name or Material Niflumic acid
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.