Learn More
MSA™ Ultravue™ Full Facepiece Assembly, Quick Disconnect
Catalog No. 19087185 Shop All MSA Products
Change view
Click to view available options
Material:
BMR, Hycar
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size
19-087-185 BMR, Hycar Large
19-087-184 BMR, Hycar Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-087-185 Supplier MSA™ Supplier No. 801452
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Facepiece without nosecup has inhalation check valve and exhalation valve that work together to avoid exhaled air from entering the mask mounted regulator and contaminating the internal regulator components.
  • Pressure-Demand Ultravue
  • Full Facepieces
  • Rubber hycar material
  • Standard 5-point head harness with quick disconnect feature
  • Without nosecup
Specifications
Certifications/Compliance NIOSH 42 CFR
Connection Type Pressure-Demand
Product Type Facepiece Assembly
Harness Style 5-strap
Includes Speaking Diaphragm
Material BMR, Hycar
Size Large
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.