Learn More
ATCC CRL-1658.2 ™ NIH/3T3.2
Supplier:  ATCC CRL1658.2
Encompass
NIH/3T3 is a fibroblast cell line that was isolated from a mouse NIH/Swiss embryo. This cell line has been subcloned from the parental material and selected specifically for contact inhibition.
Catalog No. NC3782821