Learn More
Novus Biologicals™ NIH 3T3 Hypoxic/ Normoxic Cell Lysate

Specifications
Specifications
| Preparation Method | NIH 3T3 whole cell lysate is provided as a Western blot positive control. The lysates are provided as a set of treated and untreated samples. Use 10 μL / lane for a standard mini-gel blot (1.0 mg/mL). |
| Quantity | 2 Vials |
| Host Species | Mouse |
| Content And Storage | Store at -20°C to -80°C. Avoid freeze/thaw cycles. |
| Target | NIH 3T3 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.