Learn More
Description
Nile Red is lipophilic in nature and exploits the hydrophobic properties of molecules such as lipids and plastics that fluoresce when excited with certain wavelengths, facilitating quantification and identification.
Specifications
Specifications
| CAS | 7385-67-3 |
| Molecular Weight (g/mol) | 318.37 g/mol |
| Grade | Technical |
| Color | Brown to Green |
| Quantity | 100 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.