Learn More
Nilfisk™ Safe-Pak for Nilfisk™ IVT1000 CR Vacuum
Catalog No. 19033551 Shop All Nilfisk of America Products
Change view
Click to view available options
For Use With (Equipment):
IVT1000CR Safe-Pak
Catalog No. For Use With (Equipment)
19-033-551 IVT1000CR Safe-Pak
1 options
This item is not returnable. View return policy
Catalog No. 19-033-551 Supplier Nilfisk™ Supplier No. 81303218
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protect your vacuum from contamination. The Safe-Pak™ container works by collecting hazardous dust and potent materials, and retaining them in a disposable HEPA-filtered ABS plastic container.
  • Replacement Safe-Pak HEPA-filtered Collection Container
Specifications
Product Type Replacement Safe-pak HEPA-filtered Collection Container
For Use With (Equipment) IVT1000CR Safe-Pak