Learn More
Nilfisk™ ULPA Filter for Nilfisk™ Vacuums
Catalog No. 19033572 Shop All Nilfisk of America Products
Change view
Click to view available options
For Use With (Equipment):
Lab, Cleanroom Vacuums Wet or Dry Pickup
Catalog No. For Use With (Equipment)
19-033-572 Lab, Cleanroom Vacuums Wet or Dry Pickup
1 options
This item is not returnable. View return policy
Catalog No. 19-033-572 Supplier Nilfisk™ Supplier No. L01737631
Add to Cart
Add to Cart
This item is not returnable. View return policy
Replacement ULPA exhaust filter cartridge; retains 99.999% @ 0.12 microns
Specifications
Product Type Filter Cartridge
For Use With (Equipment) Lab, Cleanroom Vacuums Wet or Dry Pickup
Filter Type ULPA