Learn More
Nilfisk™ 118EXP Explosion-Proof Vacuum
Catalog No. 19200108 Shop All Nilfisk of America Products
Change view
Click to view available options
Explosion Proof Wet Vacuum
Catalog No.
19-200-108 Explosion Proof Wet Vacuum
1 options
This item is not returnable. View return policy
Catalog No. 19-200-108 Supplier Nilfisk™ Supplier No. 1118DAXXXKT
This item is not returnable. View return policy
Avoid a major combustible dust explosion. The Nilfisk 118 EXP explosion-proof/dust ignition-proof vacuum is designed for dry material collection and features a 6.6 gallon container.
  • Made of 304 stainless steel
  • Collect and retains combustible dust within an easily removable collection chamber
  • Grounding wire and clip on filters prevent potentially dangerous static build up
  • Optional downstream HEPA and ULPA filters and bypass motor are available
Specifications
Certifications/Compliance CSA-certified for use in Class I, Group D, and Class II, Groups E*, F, and G Environments. *Additional Equipment may be necessary to meet Group E requirements. Nilfisk reserves the right to evaluate on a case-by-case basis.