Learn More
Selleck Chemical LLC Nimodipine 50mg 66085-59-4 BAY E 9736
SDP
Supplier:  Selleck Chemical LLC S174750MG
Encompass_Preferred
Nimodipine (BAY E 9736) is a dihydropyridine calcium channel blocker and an autophagy inhibitor, used in the treatment of high blood pressure. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1595