Learn More
Chemglass Life Sciences Stirrer, Magnetic, Nine Position, Dura-Mag, 230V
SDP
Supplier:  Chemglass Life Sciences CLS410009E
Encompass_Preferred
Trouble-Free Brushless MotorIntegrated Power Supply, for 100 to 220V ServiceHeavy Duty Magnets and Drive SystemTrouble-Free Non-Slip Toothed Kevlar Belt Design with Easy ReplacementStainless Steel Stirrer Plate CoverSpeed Range from 25 to 400RPM with Clockwise ControlDimensions: 16.75" W x 19.25" D x 3.5" H(front to back depth measurement includes power cord and control knob).The Chemglass Life Sciences Nine-Position magnetic stirrer is a rugged design for 24/7 laboratory or production applications.The small footprint is ideal for CO2 incubators with maximum capacity of (9) 500mL spinner vessels.Optional factory installed motion alarm available.Please note: An Optional Flask Centering Template for Corning Disposable 500mL Spinner flasks is available.Installing the drop on template ensures the Corning Disposable Spinner flasks are centered directly over the magnet for optimal stirring.
Catalog No. 50-000-83263
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.