Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninhydrin, (1,2,3-Indantrione Monohydrate), Reagents

Catalog No. 01337154
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-154 5 g
01-337-155 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-154 Supplier Reagents Holdings Llc Supplier No. C22570005C2
 Restricted Item
Add to Cart
Add to Cart

Ninhydrin, (1,2,3-Indantrione Monohydrate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.14
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Synonym 1,2,3-Triketohydrindene Monohydrate, Ninhydrine, 1,2,3-Indantrione Monohydrate
IUPAC Name 2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione
SMILES OC1(O)C(=O)C2=CC=CC=C2C1=O

Specifications

Color White to brownish-White
Packaging Glass Bottle
Quantity 5 g
Specific Gravity 0.862
Grade Reagent
Physical Form Solid
Chemical Name or Material Ninhydrin, (1,2,3-Indantrione Monohydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.