CAS RN 485-47-2
Thermo Scientific Chemicals Ninhydrin, ACS reagent
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.143 MDL Number: MFCD00003791 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N Synonym: 2,2-Dihydroxy-1,3-indanedione PubChem CID: 10236 ChEBI: CHEBI:86374 IUPAC Name: 2,2-dihydroxyindene-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Ninhydrin, 99%
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.143 MDL Number: MFCD00003791 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N Synonym: ninhydrin,ninhydrin hydrate,1,2,3-indantrione monohydrate,ninhydrine,2,2-dihydroxy-1h-indene-1,3 2h-dione,2,2-dihydroxy-1,3-indandione,triketohydrindene hydrate,1h-indene-1,3 2h-dione, 2,2-dihydroxy,1,2,3-indantrione, 2-hydrate,2,2-dihydroxyindane-1,3-dione PubChem CID: 10236 ChEBI: CHEBI:86374 IUPAC Name: 2,2-dihydroxyindene-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Thermo Scientific Chemicals Ninhydrin, ACS reagent
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.143 MDL Number: MFCD00003791 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N Synonym: 1, 2, 3-Indantrione,1, 2, 3-Triketohydrindene,2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione,1H-Indene-1, 2, 3-trione PubChem CID: 10236 ChEBI: CHEBI:86374 IUPAC Name: 2,2-dihydroxyindene-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Ninhydrin (contains Acetic Acid) Ethanol Solution [for TLC Stain], TCI America™
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.143 MDL Number: MFCD00003791 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N Synonym: ninhydrin,ninhydrin hydrate,1,2,3-indantrione monohydrate,ninhydrine,2,2-dihydroxy-1h-indene-1,3 2h-dione,2,2-dihydroxy-1,3-indandione,triketohydrindene hydrate,1h-indene-1,3 2h-dione, 2,2-dihydroxy,1,2,3-indantrione, 2-hydrate,2,2-dihydroxyindane-1,3-dione PubChem CID: 10236 ChEBI: CHEBI:86374 IUPAC Name: 2,2-dihydroxyindene-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Ninhydrin Monohydrate, MP Biomedicals™
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.14 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N IUPAC Name: 2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione SMILES: OC1(O)C(=O)C2=CC=CC=C2C1=O
Thermo Scientific Chemicals Ninhydrin, ozone friendly ready to use spray for TLC
CAS: 485-47-2 | C9H6O4 | 178.143 g/mol
Ninhydrin,, MilliporeSigma™
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.143 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N Synonym: ninhydrin,ninhydrin hydrate,1,2,3-indantrione monohydrate,ninhydrine,2,2-dihydroxy-1h-indene-1,3 2h-dione,2,2-dihydroxy-1,3-indandione,triketohydrindene hydrate,1h-indene-1,3 2h-dione, 2,2-dihydroxy,1,2,3-indantrione, 2-hydrate,2,2-dihydroxyindane-1,3-dione PubChem CID: 10236 ChEBI: CHEBI:86374 IUPAC Name: 2,2-dihydroxyindene-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Thermo Scientific Chemicals Ninhydrin, spectrophotometric grade
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.143 MDL Number: MFCD00003791 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N Synonym: 1, 2, 3-Indantrione,1, 2, 3-Triketohydrindene,2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione,1H-Indene-1, 2, 3-trione PubChem CID: 10236 ChEBI: CHEBI:86374 IUPAC Name: 2,2-dihydroxyindene-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Ninhydrin, EMD-Millipore
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.143 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N Synonym: ninhydrin,ninhydrin hydrate,1,2,3-indantrione monohydrate,ninhydrine,2,2-dihydroxy-1h-indene-1,3 2h-dione,2,2-dihydroxy-1,3-indandione,triketohydrindene hydrate,1h-indene-1,3 2h-dione, 2,2-dihydroxy,1,2,3-indantrione, 2-hydrate,2,2-dihydroxyindane-1,3-dione PubChem CID: 10236 ChEBI: CHEBI:86374 IUPAC Name: 2,2-dihydroxyindene-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
MilliporeSigma™ Ninhydrin, Calbiochem™,
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.143 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N PubChem CID: 10236 ChEBI: CHEBI:86374 IUPAC Name: 2,2-dihydroxyindene-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Ninhydrin, 0.35% (w/v) in Isopropyl Alcohol, Ricca Chemical
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): Mixture InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N PubChem CID: 10236 ChEBI: CHEBI:86374 IUPAC Name: 2,2-dihydroxyindene-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Ninhydrin 98.0+%, TCI America™
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.143 MDL Number: MFCD00003791 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N Synonym: ninhydrin,ninhydrin hydrate,1,2,3-indantrione monohydrate,ninhydrine,2,2-dihydroxy-1h-indene-1,3 2h-dione,2,2-dihydroxy-1,3-indandione,triketohydrindene hydrate,1h-indene-1,3 2h-dione, 2,2-dihydroxy,1,2,3-indantrione, 2-hydrate,2,2-dihydroxyindane-1,3-dione PubChem CID: 10236 ChEBI: CHEBI:86374 IUPAC Name: 2,2-dihydroxyindene-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
Ninhydrin, (1,2,3-Indantrione Monohydrate), Reagents
CAS: 485-47-2 Molecular Formula: C9H6O4 Molecular Weight (g/mol): 178.14 InChI Key: FEMOMIGRRWSMCU-UHFFFAOYSA-N Synonym: 1,2,3-Triketohydrindene Monohydrate, Ninhydrine, 1,2,3-Indantrione Monohydrate IUPAC Name: 2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione SMILES: OC1(O)C(=O)C2=CC=CC=C2C1=O
Sigma Aldrich Ninhydrin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich D-(+)-Neopterin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2