Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Ninhydrin, Amine Detection Reagent, Solid, 25 g
Description
- Molecular weight: 178.14 g/mol
- Chemical composition: 100% Ninhydrin
- Form: Solid
- Applications: Reacts with ammonia and primary and secondary amines to produce a blue or purple reaction product; used for development of fingermarks on porous surfaces; useful for analysis of free amino groups in proteins, peptides and amino acids; may be useful for detecting ninhydrin-reactive nitrogens in soil
- CAS number: 485-47-2
- Storage: Store at ambient temperature
Specifications
Specifications
| Target Molecule | Ammonia, Primary Amines, Secondary Amines, Amino Acids, Peptides, Proteins |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | Ammonia, Primary Amines, Secondary Amines |
| Buffer | 100% Ninhydrin |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Detection Method | Colorimetric |
| Reactivity | Ammonia, Primary amines, Secondary amines |
| Formulation | 100% Ninhydrin |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?