Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninhydrin Monohydrate, MP Biomedicals™

Catalog No. 0210245090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-102-45090 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-102-45090 Supplier MP Biomedicals, Inc Supplier No. 0210245090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ninhydrin Monohydrate is used in the detection and assay of peptides, amino acids, amino sugars and amines.

  • Ninhydrin is used for the detection of free amino groups in amino acids, peptides and proteins and in assay of peptides, amino acids, amino sugars and amines.
  • It is commonly used for the development of fingermarks on porous surfaces and is often used in comparison with other latent fingerprint techniques.
  • Studies have indicated that Ninhydrin may be useful for detecting ninhydrin-reactive nitrogens in soil for forensic applications.
  • Other studies have indicated that the color developed after reaction of Ninhydrin and amino acids can vary depending on the composition and type of amino acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.14
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
IUPAC Name 2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione
SMILES OC1(O)C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 466 to 469°F (Decomposition)
Quantity 500 g
Solubility Information 1 to 5 mg/mL at 68°F
Grade Reagent
Concentration or Composition (by Analyte or Components) 0.2% in ethanol
Physical Form Crystals
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.