Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninhydrin, 0.35% (w/v) in Isopropyl Alcohol, Ricca Chemical
SDP

Catalog No. R5309350120 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R5309350120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5309350120 Supplier Ricca Chemical Company Supplier No. R5309350120C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 485-47-2 , 67-63-0
Molecular Formula C9H6O4
Molecular Weight (g/mol) Mixture
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
PubChem CID 10236
ChEBI CHEBI:86374
IUPAC Name 2,2-dihydroxyindene-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

Specifications

Packaging Amber Glass Bottle
Color Colorless to Yellow
Quantity 120 mL
Physical Form Liquid
Percent Purity 0.5% W/V
Grade Indicator
Chemical Name or Material Ninhydrin Solution 0.35% (w/v) in Isopropyl Alcohol, In Isopropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.